Imidazo[1,2-a]pyridine-3-carbaldehyde

Supplier: Matrix Scientific

040365-500MG
101849-366EA 387.69 USD
101849-366
Imidazo[1,2-a]pyridine-3-carbaldehyde
Imidazo[1,2-a]pyridine-3-carbaldehyde

Hazard: Irritant

Formula: C₈H₆N₂O
MW: 146.1500 g/mol
MDL Number: MFCD07778354
CAS Number: 6188-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR