Tetrahydro-4H-thiopyran-4-one-1-oxide

Supplier: Matrix Scientific

Synonyms: Tetrahydro-4H-thiopyran-4-one 1-oxide

040280-500MG
101849-196EA 387.69 USD
101849-196
Tetrahydro-4H-thiopyran-4-one-1-oxide
Tetrahydro-4H-thiopyran-4-one 1-oxide

Hazard: Irritant

Formula: C₅H₈O₂S
MW: 132.1808 g/mol
MDL Number: MFCD03426617
CAS Number: 17396-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR