(5E)-5-(2,3-Dichlorobenzylidene)rhodanine

Supplier: Matrix Scientific

040186-500MG
101849-010EA 116.92 USD
101849-010
(5E)-5-(2,3-Dichlorobenzylidene)rhodanine
(5E)-5-(2,3-Dichlorobenzylidene)rhodanine

Hazard: Irritant

Formula: C₁₀H₅Cl₂NOS₂
CAS Number: 90407-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR