(5E)-5-(3-Ethoxy-2-hydroxybenzylidene)rhodanine

Supplier: Matrix Scientific

040181-500MG
101849-000EA 116.92 USD
101849-000
(5E)-5-(3-Ethoxy-2-hydroxybenzylidene)rhodanine
(5E)-5-(3-Ethoxy-2-hydroxybenzylidene)rhodanine

Hazard: Irritant

Formula: C₁₂H₁₁NO₃S₂
CAS Number: 99988-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR