(5E)-5-(2,5-Dimethylobenzylidene)rhodanine

Supplier: Matrix Scientific

040180-500MG
101848-998EA 99.38 USD
101848-998
(5E)-5-(2,5-Dimethylobenzylidene)rhodanine
(5E)-5-(2,5-Dimethylobenzylidene)rhodanine

CAS: 153567-96-5
MDL Number: MFCD04969076
Molecular Formula: C12H11NOS2
Molecular Weight: 249,36
Hazard: Irritant

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR