(5E)-5-(2-Chloro-6-fluorobenzylidene)rhodanine

Supplier: Matrix Scientific

040175-500MG
101848-988EA 116.92 USD
101848-988
(5E)-5-(2-Chloro-6-fluorobenzylidene)rhodanine
(5E)-5-(2-Chloro-6-fluorobenzylidene)rhodanine

Hazard: Irritant

Formula: C₁₀H₅ClFNOS₂
CAS Number: 217316-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR