(5E)-5-(2,6-Dichlorobenzylidene)rhodanine

Supplier: Matrix Scientific

040173-500MG
101848-984EA 99.38 USD
101848-984
(5E)-5-(2,6-Dichlorobenzylidene)rhodanine
(5E)-5-(2,6-Dichlorobenzylidene)rhodanine

CAS: 65562-49-4
MDL Number: MFCD04969007
Molecular Formula: C10H5Cl2NOS2
Molecular Weight: 290,19
Hazard: Irritant

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR