(5E)-5-(4-Chlorobenzylidene)rhodanine

Supplier: Matrix Scientific

040170-500MG
101848-978EA 116.92 USD
101848-978
(5E)-5-(4-Chlorobenzylidene)rhodanine
(5E)-5-(4-Chlorobenzylidene)rhodanine

Hazard: Irritant

Formula: C₁₀H₆ClNOS₂
CAS Number: 81154-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR