(5E)-5-(3-Ethoxy-4-hydroxybenzylidene)rhodanine

Supplier: Matrix Scientific

040162-500MG
101848-962EA 116.92 USD
101848-962
(5E)-5-(3-Ethoxy-4-hydroxybenzylidene)rhodanine
(5E)-5-(3-Ethoxy-4-hydroxybenzylidene)rhodanine

Hazard: Irritant

Formula: C₁₂H₁₁NO₃S₂
CAS Number: 6322-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR