(5E)-5-(3,4-Dimethoxybenzylidene)rhodanine

Supplier: Matrix Scientific

040159-500MG
101848-956EA 87.4 USD
101848-956
(5E)-5-(3,4-Dimethoxybenzylidene)rhodanine
(5E)-5-(3,4-Dimethoxybenzylidene)rhodanine

Hazard: Irritant

Formula: C₁₂H₁₁NO₃S₂
CAS Number: 6326-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR