(5E)-2-Mercapto-5-(4-(trifluoromethyl)benzylidene)-1,3-thiazol-4(5H)-one

Supplier: Matrix Scientific

040148-500MG
101848-934EA 116.92 USD
101848-934
(5E)-2-Mercapto-5-(4-(trifluoromethyl)benzylidene)-1,3-thiazol-4(5H)-one
(5E)-2-Mercapto-5-(4-(trifluoromethyl)benzylidene)-1,3-thiazol-4(5H)-one

Hazard: Irritant

Formula: C₁₁H₆F₃NOS₂
CAS Number: 99460-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR