(5E)-5-(4-tert-Butylbenzylidene)rhodanine

Supplier: Matrix Scientific

Synonyms: (5E)-5-(4-tert-Butylbenzylidene)-2-mercapto-1,3-thiazol-4(5H)-one

040145-500MG
101848-928EA 87.4 USD
101848-928
(5E)-5-(4-tert-Butylbenzylidene)rhodanine
(5E)-5-(4-tert-Butylbenzylidene)rhodanine

Hazard: Irritant

Formula: C₁₄H₁₅NOS₂
CAS Number: 312607-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR