3-Amino-5-bromo-1H-indazole

Supplier: Matrix Scientific

Synonyms: 5-Bromo-1H-indazol-3-amine

037009-500MG
101842-500EA 32.31 USD
101842-500
3-Amino-5-bromo-1H-indazole
3-Amino-5-bromo-1H-indazole

Hazard: Irritant

Formula: C₇H₆BrN₃
MW: 212.05 g/mol
MDL Number: MFCD03426696
CAS Number: 61272-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR