2-(Methylsulfonyl)acetophenone

Supplier: Matrix Scientific

Synonyms: 2-(Methylsulfonyl)-1-phenylethanone2-(Methylsulphonyl)-1-phenylethanone

036985-1G
101842-462EA 203.08 USD
101842-462
2-(Methylsulfonyl)acetophenone
2-(Methylsulphonyl)acetophenone

Hazard: Irritant

Formula: C₉H₁₀O₃S
Melting Pt: 110 °C
MDL Number: MFCD00025078
CAS Number: 3708-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR