2-Ethoxy-5-nitrobenzonitrile

Supplier: Matrix Scientific

036951-500MG
101842-398EA 146.15 USD
101842-398
2-Ethoxy-5-nitrobenzonitrile
2-Ethoxy-5-nitrobenzonitrile

Hazard: Irritant

Formula: C₉H₈N₂O₃
MW: 192.1700 g/mol
MDL Number: MFCD02256084
CAS Number: 29669-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR