3-Chloro-6-(cyclopropylamino)pyridazine

Supplier: Matrix Scientific

Synonyms: 6-Chloro-N-cyclopropylpyridazin-3-amine

036836-500MG
101842-176EA 193.85 USD
101842-176
3-Chloro-6-(cyclopropylamino)pyridazine
3-Chloro-6-(cyclopropylamino)pyridazine

Hazard: Irritant

Formula: C₇H₈ClN₃
MW: 169.6100 g/mol
MDL Number: MFCD09800959
CAS Number: 941294-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR