2-Ethoxynicotinonitrile

Supplier: Matrix Scientific

036797-500MG
101842-102EA 55.38 USD
101842-102
2-Ethoxynicotinonitrile
2-Ethoxynicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile

Hazard: Irritant

Formula: C₇H₆N₂O
MDL Number: MFCD08436064
CAS Number: 14248-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR