2-Methoxynicotinonitrile

Supplier: Matrix Scientific

Synonyms: 3-Cyano-2-methoxypyridine

036795-500MG
101842-098EA 115 USD
101842-098
2-Methoxynicotinonitrile
2-Methoxynicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile

Hazard: Irritant

Formula: C₇H₆N₂O
MW: 134.14 g/mol
MDL Number: MFCD07363804
CAS Number: 7254-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR