5-Amino-2-methoxybenzonitrile

Supplier: Matrix Scientific

036762-500MG
101842-028EA 387.69 USD
101842-028
5-Amino-2-methoxybenzonitrile
5-Amino-2-methoxybenzonitrile

Hazard: Irritant

Formula: C₈H₈N₂O
MW: 148.1619 g/mol
MDL Number: MFCD00723841
CAS Number: 214623-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR