5-Amino-2-ethoxybenzonitrile

Supplier: Matrix Scientific

036755-500MG
101842-014EA 362.25 USD
101842-014
5-Amino-2-ethoxybenzonitrile
5-Amino-2-ethoxybenzonitrile

Hazard: Irritant

Formula: C₉H₁₀N₂O
MDL Number: MFCD10686562
CAS Number: 1020046-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR