N-(2-Hydroxyethyl)-2-cyano-4-nitroaniline

Supplier: Matrix Scientific

Synonyms: 2-Cyano-N-(2-hydroxyethyl)-4-nitroaniline2-(2-Hydroxyethylamino)-5-nitrobenzonitrile

036690-500MG
101841-886EA 193.85 USD
101841-886
N-(2-Hydroxyethyl)-2-cyano-4-nitroaniline
N-(2-Hydroxyethyl)-2-cyano-4-nitroaniline

CAS: 63989-40-2
MDL Number: MFCD00051934
Molecular Formula: C9H9N3O3
Molecular Weight: 207,19
Hazard: Irritant

Formula: C₉H₉N₃O₃
MW: 207.19 g/mol
MDL Number: MFCD00051934
CAS Number: 63989-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR