2-(Cyclopropylamino)nicotinonitrile

Supplier: Matrix Scientific

036648-500MG
101841-802EA 193.85 USD
101841-802
2-(Cyclopropylamino)nicotinonitrile
2-(Cyclopropylamino)nicotinonitrile

Hazard: Irritant

Formula: C₉H₉N₃
MDL Number: MFCD09455268
CAS Number: 52583-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR