(5-Methyl-1H-pyrazol-1-yl)acetic acid

Supplier: Matrix Scientific

036599-500MG
101841-704EA 166.75 USD
101841-704
(5-Methyl-1H-pyrazol-1-yl)acetic acid
(5-Methyl-1H-pyrazol-1-yl)acetic acid

Hazard: Irritant

Formula: C₆H₈N₂O₂
MDL Number: MFCD03419599
CAS Number: 180741-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR