1-(2-Cyanoethyl)-2-methylimidazole

Supplier: Matrix Scientific

Synonyms: 3-(2-Methyl-1H-imidazol-1-yl)propanenitrile3-(2-Methyl-1-imidazolyl)propionitrile

036571-500MG
101841-648EA 387.69 USD
101841-648
1-(2-Cyanoethyl)-2-methylimidazole
1-(2-Cyanoethyl)-2-methylimidazole

Hazard: Irritant

Formula: C₇H₉N₃
MW: 135.17 g/mol
MDL Number: MFCD00047049
CAS Number: 23996-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR