2-(1H-Imidazol-1-yl)acetonitrile

Supplier: Matrix Scientific

Synonyms: 1-(Cyanomethyl)imidazole(1-Imidazolyl)acetonitrile

036568-500MG
101841-642EA 387.69 USD
101841-642
2-(1H-Imidazol-1-yl)acetonitrile
2-(1H-Imidazol-1-yl)acetonitrile

Hazard: Irritant

Formula: C₅H₅N₃
MW: 107.12 g/mol
MDL Number: MFCD06421433
CAS Number: 98873-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR