Morpholinoacetonitrile
Supplier: Matrix Scientific
Synonyms:
N-Cyanomethylmorpholine
036566-1G
036566-5G
101841-636EA
27.69
USD
101841-636
101841-638
Morpholinoacetonitrile
Morpholinoacetonitrile
CAS: 5807-02-3
MDL Number: MFCD00023372
Molecular Formula: C6H10N2O
Molecular Weight: 126,16
Boiling Point: 78°C/2 Torr
Hazard: Irritant
Formula:
C₆H₁₀N₂O MW: 126.16 g/mol Boiling Pt: 78 °C (2 Torr) Melting Pt: 60…61 °C |
MDL Number:
MFCD00023372 CAS Number: 5807-02-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...