4-Bromo-1-ethyl-3,5-dimethyl-1H-pyrazole

Supplier: Matrix Scientific

036448-500MG
101841-386EA 387.69 USD
101841-386
4-Bromo-1-ethyl-3,5-dimethyl-1H-pyrazole
4-Bromo-1-ethyl-3,5-dimethyl-1H-pyrazole

Hazard: Irritant

Formula: C₇H₁₁BrN₂
MDL Number: MFCD11054017
CAS Number: 51108-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR