4-Bromo-1-ethyl-3-methyl-1H-pyrazole

Supplier: Matrix Scientific

036440-500MG
101841-368EA 190.77 USD
101841-368
4-Bromo-1-ethyl-3-methyl-1H-pyrazole
4-Bromo-1-ethyl-3-methyl-1H-pyrazole

Hazard: Irritant

Formula: C₆H₉BrN₂
MDL Number: MFCD02655588
CAS Number: 519018-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR