4-Chloro-1,3-dimethyl-1H-pyrazole

Supplier: Matrix Scientific

036436-500MG
101841-360EA 387.69 USD
101841-360
4-Chloro-1,3-dimethyl-1H-pyrazole
4-Chloro-1,3-dimethyl-1H-pyrazole

Hazard: Irritant

Formula: C₅H₇ClN₂
MDL Number: MFCD09743376
CAS Number: 15878-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR