4-Bromo-1-methyl-1H-pyrazole ≥97%

Supplier: Matrix Scientific

036427-25G
101841-342EA 66.15 USD
101841-342
4-Bromo-1-methyl-1H-pyrazole ≥97%
4-Bromo-1-methyl-1H-pyrazole

Hazard: Irritant

Formula: C₄H₅BrN₂
MW: 161.00 g/mol
MDL Number: MFCD02179565
CAS Number: 15803-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR