5-(Chloromethyl)-3-(2-methylphenyl)-1,2,4-oxadiazole

Supplier: Matrix Scientific

036236-500MG
101840-956EA 146.15 USD
101840-956
5-(Chloromethyl)-3-(2-methylphenyl)-1,2,4-oxadiazole
5-(Chloromethyl)-3-(2-methylphenyl)-1,2,4-oxadiazole

Hazard: Irritant

Formula: C₁₀H₉ClN₂O
MDL Number: MFCD06358646
CAS Number: 60580-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR