3-(5-Methyl-1,2,4-oxadiazol-3-yl)benzaldehyde

Supplier: Matrix Scientific

036218-500MG
101840-920EA 440 USD
101840-920
3-(5-Methyl-1,2,4-oxadiazol-3-yl)benzaldehyde
3-(5-Methyl-1,2,4-oxadiazol-3-yl)benzaldehyde

Hazard: Irritant

Formula: C₁₀H₈N₂O₂
MDL Number: MFCD07368545
CAS Number: 852180-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR