3-{5-[(Isobutylamino)methyl]-1,2,4-oxadiazol-3-yl}benzamide

Supplier: Matrix Scientific

036213-500MG
101840-910EA 484.62 USD
101840-910
3-{5-[(Isobutylamino)methyl]-1,2,4-oxadiazol-3-yl}benzamide
3-{5-[(Isobutylamino)methyl]-1,2,4-oxadiazol-3-yl}benzamide

Hazard: Irritant

Formula: C₁₄H₁₈N₄O₂
MDL Number: MFCD12026810
CAS Number: 1119450-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR