3-(Aminomethyl)-N,N-dimethyl-1,2,4-oxadiazole-5-carboxamide hydrochloride

Supplier: Matrix Scientific

036205-500MG
101840-894EA 484.62 USD
101840-894
3-(Aminomethyl)-N,N-dimethyl-1,2,4-oxadiazole-5-carboxamide hydrochloride
3-(Aminomethyl)-N,N-dimethyl-1,2,4-oxadiazole-5-carboxamide hydrochloride

Hazard: Irritant

Formula: C₆H₁₁ClN₄O₂
MDL Number: MFCD12026802
CAS Number: 1185299-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR