3-(Aminomethyl)-N-(2-amino-2-oxoethyl)-1,2,4-oxadiazole-5-carboxamide

Supplier: Matrix Scientific

036203-500MG
101840-890EA 484.62 USD
101840-890
3-(Aminomethyl)-N-(2-amino-2-oxoethyl)-1,2,4-oxadiazole-5-carboxamide
3-(Aminomethyl)-N-(2-amino-2-oxoethyl)-1,2,4-oxadiazole-5-carboxamide

Hazard: Irritant

Formula: C₆H₉N₅O₃
MDL Number: MFCD12026801
CAS Number: 1119452-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR