(5-Cyclopentyl-1,2,4-oxadiazol-3-yl)methylamine

Supplier: Matrix Scientific

036189-500MG
101840-860EA 307.69 USD
101840-860
(5-Cyclopentyl-1,2,4-oxadiazol-3-yl)methylamine
(5-Cyclopentyl-1,2,4-oxadiazol-3-yl)methylamine

Hazard: Irritant

Formula: C₈H₁₃N₃O
MDL Number: MFCD08691632
CAS Number: 915925-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR