2-(2-Pyrazin-2-yl-1,3-thiazol-4-yl)ethanamine

Supplier: Matrix Scientific

036171-500MG
101840-824EA 290.77 USD
101840-824
2-(2-Pyrazin-2-yl-1,3-thiazol-4-yl)ethanamine
2-(2-Pyrazin-2-yl-1,3-thiazol-4-yl)ethanamine

Hazard: Irritant

Formula: C₉H₁₀N₄S
MDL Number: MFCD11053908
CAS Number: 1060817-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR