Methyl-2-methyl-3-(1H-pyrazol-1-yl)propanoate
Supplier: Matrix Scientific
036162-500MG
101840-806EA
218.46
USD
101840-806
Methyl-2-methyl-3-(1H-pyrazol-1-yl)propanoate
Methyl-2-methyl-3-(1H-pyrazol-1-yl)propanoate
Hazard: Irritant
Formula:
C₈H₁₂N₂O₂ Boiling Pt: 118 °C (11 Torr) |
MDL Number:
MFCD09473571 CAS Number: 90197-40-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...