Ethyl [(6,7-difluoro-1,3-benzoxazol-2-yl)thio]acetate

Supplier: Matrix Scientific

036152-500MG
101840-786EA 290.77 USD
101840-786
Ethyl [(6,7-difluoro-1,3-benzoxazol-2-yl)thio]acetate
Ethyl [(6,7-difluoro-1,3-benzoxazol-2-yl)thio]acetate

Hazard: Irritant

Formula: C₁₁H₉F₂NO₃S
MDL Number: MFCD11696463
CAS Number: 1071369-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR