6-Chloro-2-(methylthio)-1,3-benzoxazole

Supplier: Matrix Scientific

036144-500MG
101840-770EA 193.85 USD
101840-770
6-Chloro-2-(methylthio)-1,3-benzoxazole
6-Chloro-2-(methylthio)-1,3-benzoxazole

Hazard: Irritant

Formula: C₈H₆ClNOS
MDL Number: MFCD11696461
CAS Number: 1071296-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR