Ethyl-3-[ethyl(methyl)amino]butanoate

Supplier: Matrix Scientific

036131-500MG
101840-744EA 387.69 USD
101840-744
Ethyl-3-[ethyl(methyl)amino]butanoate
Ethyl-3-[ethyl(methyl)amino]butanoate

Hazard: Irritant

Formula: C₉H₁₉NO₂
MDL Number: MFCD11696459
CAS Number: 1071401-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR