Ethyl-3-(dimethylamino)butanoate

Supplier: Matrix Scientific

036129-500MG
101840-740EA 290.77 USD
101840-740
Ethyl-3-(dimethylamino)butanoate
Ethyl-3-(dimethylamino)butanoate

Hazard: Irritant

Formula: C₈H₁₇NO₂
MDL Number: MFCD11696458
CAS Number: 85118-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR