3-(Dimethylamino)butan-1-ol

Supplier: Matrix Scientific

036128-500MG
101840-738EA 423.08 USD
101840-738
3-(Dimethylamino)butan-1-ol
3-(Dimethylamino)butan-1-ol

Hazard: Irritant

Formula: C₆H₁₅NO
MDL Number: MFCD09971253
CAS Number: 2893-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR