2-(4-Methylphenyl)-1,2,4-benzotriazin-3(2H)-one

Supplier: Matrix Scientific

036120-500MG
101840-722EA 484.62 USD
101840-722
2-(4-Methylphenyl)-1,2,4-benzotriazin-3(2H)-one
2-(4-Methylphenyl)-1,2,4-benzotriazin-3(2H)-one

Hazard: Irritant

Formula: C₁₄H₁₁N₃O
MDL Number: MFCD11696455
CAS Number: 1071296-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR