3-(1H-Pyrazol-1-yl)butan-1-ol

Supplier: Matrix Scientific

036118-500MG
101840-720EA 290.77 USD
101840-720
3-(1H-Pyrazol-1-yl)butan-1-ol
3-(1H-Pyrazol-1-yl)butan-1-ol

Hazard: Irritant

Formula: C₇H₁₂N₂O
MDL Number: MFCD08701090
CAS Number: 1007517-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR