2-(1H-Pyrazol-1-yl)propan-1-ol

Supplier: Matrix Scientific

036113-500MG
101840-710EA 387.69 USD
101840-710
2-(1H-Pyrazol-1-yl)propan-1-ol
2-(1H-Pyrazol-1-yl)propan-1-ol

Hazard: Irritant

Formula: C₆H₁₀N₂O
MDL Number: MFCD09864511
CAS Number: 191725-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR