[3-(4-Fluorophenyl)-1H-pyrazol-4-yl]methanol

Supplier: Matrix Scientific

036105-500MG
101840-694EA 323.08 USD
101840-694
[3-(4-Fluorophenyl)-1H-pyrazol-4-yl]methanol
[3-(4-Fluorophenyl)-1H-pyrazol-4-yl]methanol

Hazard: Irritant

Formula: C₁₀H₉FN₂O
MDL Number: MFCD06203022
CAS Number: 1017783-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR