3-[(2-Methoxyethyl)(methyl)amino]propanoic acid hydrochloride

Supplier: Matrix Scientific

036071-500MG
101840-628EA 116.92 USD
101840-628
3-[(2-Methoxyethyl)(methyl)amino]propanoic acid hydrochloride
3-[(2-Methoxyethyl)(methyl)amino]propanoic acid hydrochloride

Hazard: Irritant

Formula: C₇H₁₆ClNO₃
MDL Number: MFCD11696440
CAS Number: 915922-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR