Methyl-3-piperidin-1-ylpropanoate

Supplier: Matrix Scientific

036070-500MG
101840-626EA 290.77 USD
101840-626
Methyl-3-piperidin-1-ylpropanoate
Methyl-3-piperidin-1-ylpropanoate

Hazard: Irritant

Formula: C₉H₁₇NO₂
MDL Number: MFCD00461067
CAS Number: 23573-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR