1H-Benzimidazole-2-sulfonic acid

Supplier: Matrix Scientific

Synonyms: 1H-Benzo[d]imidazole-2-sulfonic acid1H-Benzo[d]imidazole-2-sulphonic acid

036059-500MG
101840-604EA 387.69 USD
101840-604
1H-Benzimidazole-2-sulfonic acid
1H-Benzimidazole-2-sulphonic acid

Hazard: Irritant

Formula: C₇H₆N₂O₃S
MW: 198.1991 g/mol
MDL Number: MFCD00100972
CAS Number: 40828-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR